Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-leucine 98.0+%, TCI America™
SDP

Catalog No. p-7130034 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 103478-62-2
Molecular Formula C22H25NO4
Molecular Weight (g/mol) 367.45
MDL Number MFCD00151933
InChI Key BUJQSIPFDWLNDC-GNLPSFAGNA-N
Synonym fmoc-n-methyl-l-leucine, fmoc-n-me-leu-oh, n-fmoc-n-methyl-l-leucine, fmoc-meleu-oh, l-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-4-methyl-pentanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-4-methylpentanoic acid, pubchem13155, fmoc-n-a-methyl-l-leucine, n-methyl-n-fmoc-l-leu-oh
PubChem CID 7015835
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-4-methylpentanoic acid
SMILES CC(C)C[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 1 g
Formula Weight 367.45
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-leucine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.