Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-D-Galactosamine, MP Biomedicals™

Catalog No. p-7083073 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 mg
500mg

A component of many O-linked and N-linked glycan structures. N-Acetyl-D-Galactosamine, MP Biomedicals is an aminosugar that is the initial O-linked sugar to many serine and threonine residues in protein glycosylations.

Chemical Identifiers

CAS 14215-68-0
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.209
InChI Key OVRNDRQMDRJTHS-JAJWTYFOSA-N
Synonym n-acetyl-d-galactosamine, galnac, 2-acetamido-2-deoxy-d-galactopyranose, d-galnac, 2-acetamido-2-deoxy-d-galactose, n-acetyl-d-chondrosamine, n-3r,4r,5r,6r-2,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-3-yl acetamide, n-acetylgalactosamine, acetylgalactosamine, n-acetyl-galactosamine
PubChem CID 440552
ChEBI CHEBI:28497
IUPAC Name N-[(2R,3R,4R,5R,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)NC1C(C(C(OC1O)CO)O)O

Specifications

Melting Point 160°C to 161°C (literature)
Quantity 100 mg
Solubility Information Soluble in water (100mg/mL - clear, colorless solution).
Optical Rotation +84–89° (c=1, water)
Formula Weight 221.21
Physical Form Powder
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.