Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-4-(2-hydroxyethyl)piperidine, 97%, Thermo Scientific™

Catalog No. AC296480010
Click to view available options
Quantity:
Packaging:

 

Chemical Identifiers

CAS

76876-70-5

Molecular Formula

C14H21NO

Molecular Weight (g/mol)

219.33

MDL Number

MFCD00800492

InChI Key

FBFPTWSKNJHCGT-UHFFFAOYSA-N

Synonym

2-1-benzylpiperidin-4-yl ethanol, n-benzyl-4-2-hydroxyethyl piperidine, 1-benzyl-4-2-hydroxyethyl piperdine, 1-benzyl-4-2-hydroxyethyl piperidine, 2-1-benzyl-4-piperidyl ethan-1-ol, 1-benzylpiperidine-4-ethanol, 1-phenylmethyl-4-piperidineethanol, 4-piperidineethanol,1-phenylmethyl, 2-1-benzylpiperidin-4-yl ethan-1-ol, n-benzyl-4-2-hydroxyethyl-piperidine

PubChem CID

2725029

IUPAC Name

2-(1-benzylpiperidin-4-yl)ethanol

SMILES

C1CN(CCC1CCO)CC2=CC=CC=C2

Specifications

CAS Min %

96.0

CAS Max %

100.0

Color

Yellow to Brown

Infrared Spectrum

Authentic

Assay Percent Range

96% min. (GC)

Packaging

Glass bottle

Refractive Index

1.5370 to 1.5400

Quantity

1 g

Formula Weight

219.33

Percent Purity

97%

Physical Form

Viscous Liquid

Chemical Name or Material

N-Benzyl-4-(2-hydroxyethyl)piperidine, 97%

Hazard Category

GHS Signal Word: Warning

Hazard Statement

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Precautionary Statement

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.