Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[Bis(methylthio)methylene]-p-toluenesulfonamide 97.0+%, TCI America™
SDP

Catalog No. B13895G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 2651-15-2
Molecular Formula C10H13NO2S3
Molecular Weight (g/mol) 275.40
MDL Number MFCD00144854
InChI Key OWIPGZGAUSIOAX-UHFFFAOYSA-N
Synonym N-[Bis(methylthio)methylene]-N-tosylamine
PubChem CID 319370
IUPAC Name N-[bis(methylsulfanyl)methylidene]-4-methylbenzene-1-sulfonamide
SMILES CSC(SC)=NS(=O)(=O)C1=CC=C(C)C=C1

Specifications

Melting Point 111°C
Color White-Yellow
Quantity 5 g
Formula Weight 275.40
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-[Bis(methylthio)methylene]-p-toluenesulfonamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.