Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-Hexahydro-1H-azepin-4-one, 97%, Thermo Scientific Chemicals

Catalog No. p-4228413
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 188975-88-4
Molecular Formula C11H19NO3
Molecular Weight (g/mol) 213.28
MDL Number MFCD03788435
InChI Key PMLBUVZPRKXMOX-UHFFFAOYSA-N
Synonym n-boc-hexahydro-1h-azepin-4-one, 1-boc-hexahydro-1h-azepin-4-one, 4-oxoazepane-1-carboxylic acid tert-butyl ester, 1-boc-homopiperazin-4-one, 1-boc-4-azepanone, n-boc-hexahydro-1h-azepine-4-one, tert-butyl hexahydro-4-oxo-1h-azepine-1-carboxylate, 1-tert-butoxycarbonyl-4-oxoazepane, 4-oxo-azepane-1-carboxylic acid tert-butyl ester, 1,1-dimethylethyl 4-oxohexahydro-1h-azepine-1-carboxylate
PubChem CID 1512679
IUPAC Name tert-butyl 4-oxoazepane-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(=O)CC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 213.28
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material N-BOC-Hexahydro-1H-azepin-4-one
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.