Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butanol, 7.5% v/v, Fisher Chemical™
SDP

Catalog No. p-213103 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 71-36-3, 7732-18-5, 497-19-8, 144-55-8
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

CAS Min % 92.0, 5.54, 8.0E-4, 7.0E-4
CAS Max % 96.0, 5.77, 8.0E-4, 7.0E-4
Quantity 1 L
Packaging Amber Glass Bottle
Chemical Name or Material n-Butanol, 7.5% v/v
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.