Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Hydroxymethylphthalimide 98.0+%, TCI America™
SDP

Catalog No. p-7130658 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 118-29-6
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.159
MDL Number MFCD00005899
InChI Key MNSGOOCAMMSKGI-UHFFFAOYSA-N
Synonym n-hydroxymethyl phthalimide, n-hydroxymethylphthalimide, 2-hydroxymethyl isoindoline-1,3-dione, phthalimidomethanol, phthalimidomethyl alcohol, n-methylolphthalimide, oxymethyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-hydroxymethyl, hydroxymethylphthalimide, methanol, phthalimido
PubChem CID 8354
ChEBI CHEBI:38816
IUPAC Name 2-(hydroxymethyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CO

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 25 g
Formula Weight 177.16
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Hydroxymethylphthalimide
RTECSNumber TI5270000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.