Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-2-morpholino-5-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™

Catalog No. CC57846DA
Click to view available options
Quantity:

Chemical Identifiers

CAS

886851-52-1

Molecular Formula

C13H17F3N2O

Molecular Weight (g/mol)

274.29

MDL Number

MFCD09025891

InChI Key

UAZNBBVDDCKGES-UHFFFAOYSA-N

Synonym

n-methyl-2-morpholino-5-trifluoromethyl benzylamine, methyl 2-morpholin-4-yl-5-trifluoromethyl phenyl methyl amine, n-methyl-n-2-morpholin-4-yl-5-trifluoromethyl benzyl amine, benzenemethanamine,n-methyl-2-4-morpholinyl-5-trifluoromethyl, n-methyl-1-2-morpholino-5-trifluoromethyl phenyl methanamine, n-methyl-1-2-morpholin-4-yl-5-trifluoromethyl phenyl methanamine

PubChem CID

18525888

IUPAC Name

N-methyl-1-[2-morpholin-4-yl-5-(trifluoromethyl)phenyl]methanamine

SMILES

CNCC1=C(C=CC(=C1)C(F)(F)F)N1CCOCC1

Specifications

CAS Min %

96.5

CAS Max %

100.0

Assay Percent Range

97%

Quantity

1 g

Formula Weight

274.28

Percent Purity

97%

Chemical Name or Material

N-Methyl-2-morpholino-5-(trifluoromethyl)benzylamine

Hazard Statement

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

Precautionary Statement

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.