Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N-(4-pyrrolidin-1-ylbenzyl)amine, 90%, Thermo Scientific™

Catalog No. p-3175722
Click to view available options
Quantity:
1 g
10 g

Chemical Identifiers

CAS 823188-79-0
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD07686040
InChI Key DJSIBXGPJNBMBD-UHFFFAOYSA-N
Synonym n-methyl-n-4-pyrrolidin-1-ylbenzyl amine, methyl 4-pyrrolidin-1-yl phenyl methyl amine, n-methyl-1-4-pyrrolidin-1-yl phenyl methanamine, n-methyl-1-4-pyrrolidin-1-ylphenyl methanamine, n-methyl 4-1-pyrrolidinyl phenyl methanamine, benzenemethanamine,n-methyl-4-1-pyrrolidinyl, benzenemethanamine, n-methyl-4-1-pyrrolidinyl
PubChem CID 7064073
IUPAC Name methyl({[4-(pyrrolidin-1-yl)phenyl]methyl})amine
SMILES CNCC1=CC=C(C=C1)N1CCCC1

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 190.29
Percent Purity 90%
Chemical Name or Material N-Methyl-N-(4-pyrrolidin-1-ylbenzyl)amine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.