Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylpyrrolidinone, Fisher BioReagents

Catalog No. p-27176 Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L
19 L
Packaging:
Glass Bottle
NOWPak™ I (Plastic Exterior)
This item is not returnable. View return policy
This item is not returnable. View return policy

For automated oligonucleotide synthesis

N-Methylpyrrolidinone is used in automated oligonucleotide synthesis.

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

pH 7.7 to 8.0
Melting Point -24°C
Absorbance 0.02 AU max. at 400nm, 0.3 AU max. at 300nm, 0.7 AU max. at 270nm, 1.0 AU max. at 265nm
Color Colorless
Density 1.030g/cm³
Boiling Point 202°C
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range (by GC): ≥99.8%
Titratable Base 0.001mEq/g max.
Vapor Pressure 0.7mbar at 25°C
Water 0.05% max.
Merck Index 15, 6190
Formula Weight 99.13
Percent Purity ≥99.8%
Chemical Name or Material N-Methylpyrrolidinone
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Combustible liquid. Developmental hazard. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Obtain medical attention.

NFPA

Health:2
Flammability:2
Instability:0
Recommended Storage Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.