Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylacrylamide, 99%, stabilized with 500 ppm MEHQ, Thermo Scientific Chemicals

Catalog No. p-4228616
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2680-03-7
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
InChI Key YLGYACDQVQQZSW-UHFFFAOYSA-N
Synonym n,n-dimethylacrylamide, 2-propenamide, n,n-dimethyl, acrylamide, n,n-dimethyl, acylamide, n,n-dimethyl, n,n-dimethyl-acrylamide, n,n-dimethyl-2-propenamide, dimethylamid kyseliny akrylove, nn-dimethylacrylamide, unii-as46jk7q6i, propenamide, n,n-dimethyl
PubChem CID 17587
IUPAC Name N,N-dimethylprop-2-enamide
SMILES CN(C)C(=O)C=C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9620g/mL
Boiling Point 80.0°C (20.0 mmHg)
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4720 to 1.4740 (20°C, 589nm)
Quantity 100 mL
Specific Gravity 0.962
Solubility Information Solubility in water: slightly soluble.
Stabilizer 500ppm (approximately)
Formula Weight 99.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylacrylamide, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
EINECSNumber 220-237-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.