Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethylformamide, Biotech Grade, ≥99.9%, Honeywell Burdick & Jackson™

Catalog No. p-7120614 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1L
2 L
6 x 1 L
Packaging:
Glass Bottle

Biotech Grade,≥99.9%

  • Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
  • As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
  • Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Linear Formula HCON(CH3)2
UN Number UN2265
Beilstein 605365
Formula Weight 73.09g/mol
Percent Purity ≥99.9%
Density 0.944g/mL
Explosion Limit 0.152
Ignition Point 833°F
Odor Weak
Packaging Glass Bottle
Vapor Pressure 2.7mmHg (20°C)
Vapor Density 2.5 (vs air)
Flash Point 58°C (136.4°F)
Physical State Liquid
Boiling Point 152°C to 153°C
Color Colorless
Melting Point -61°C
pH 7 (at 20°C)
Quantity 1L
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
Hazard Statement P201-P280-P305 + P351 + P338-P308 + P313
DOTInformation 3/PGIII
RTECSNumber 200-679-5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.