Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N′,N′′,N′′′-Tetraacetylchitotetraose 98.0+%, TCI America™
SDP

Catalog No. T291010MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

2706-65-2

Molecular Formula

C32H54N4O21

Molecular Weight (g/mol)

830.791

MDL Number

MFCD00136030

InChI Key

VDENSBHPZQKGGL-YCAJNKKBSA-N

Synonym

Tetra-N-acetylchitotetraose, GlcNAc beta(1-4)GlcNAc beta(1-4)GlcNAc beta(1-4)GlcNAc

PubChem CID

131698500

IUPAC Name

N-[(2S,3R,4R,5S,6R)-2-[(2R,3S,4R,5R,6S)-5-acetamido-6-[(2R,3S,4R,5R,6S)-5-acetamido-6-[(2R,4R,5R)-5-acetamido-1,2,4-trihydroxy-6-oxohexan-3-yl]oxy-4-hydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4-hydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-6-(hydroxyme

SMILES

CC(=O)NC1C(C(C(OC1OC2C(OC(C(C2O)NC(=O)C)OC3C(OC(C(C3O)NC(=O)C)OC(C(CO)O)C(C(C=O)NC(=O)C)O)CO)CO)CO)O)O

Specifications

Melting Point

300°C

Color

White

Quantity

10 mg

Formula Weight

830.79

Percent Purity

≥98.0% (HPLC)

Physical Form

Crystalline Powder

Chemical Name or Material

N,N′,N′′,N′′′-Tetraacetylchitotetraose

TSCA

No

Recommended Storage

Freezer

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.