Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Nitrosoheptamethyleneimine 99.0+%, TCI America™
SDP

Catalog No. p-7133813 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 20917-49-1
Molecular Formula C7H14N2O
Molecular Weight (g/mol) 142.20
MDL Number MFCD00059811
InChI Key RCFKQVLHWYOSFF-UHFFFAOYSA-N
Synonym Heptamethylenenitrosamine
PubChem CID 30360
IUPAC Name 1-nitrosoazocane
SMILES O=NN1CCCCCCC1

Specifications

Color Yellow
Quantity 100 mg
UN Number 2811
Formula Weight 142.20
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material N-Nitrosoheptamethyleneimine
RTECSNumber CN4900000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.