Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl 6-Biotinamidohexanoate 97.0+%, TCI America™
SDP

Catalog No. p-7135909 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
20 mg
100 mg

Chemical Identifiers

CAS 72040-63-2
Molecular Formula C20H30N4O6S
Molecular Weight (g/mol) 454.542
MDL Number MFCD00065502
InChI Key UVGHPGOONBRLCX-NJSLBKSFSA-N
Synonym 6-Biotinamidohexanoic Acid N-Succinimidyl Ester, N-[5-(N-Succinimidyloxycarbonyl)pentyl]biotinamide, N-Biotinyl-6-aminohexanoic Acid Succinimidyl Ester
PubChem CID 83874
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 6-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]hexanoate
SMILES C1CC(=O)N(C1=O)OC(=O)CCCCCNC(=O)CCCCC2C3C(CS2)NC(=O)N3

Specifications

Melting Point 170°C
Color White
Quantity 20 mg
Formula Weight 454.54
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 6-Biotinamidohexanoate
TSCA No
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.