Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-D-phenylalaninol 97.0+%, TCI America™
SDP

Catalog No. p-7119387 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 106454-69-7
Molecular Formula C14H21NO3
Molecular Weight (g/mol) 251.33
MDL Number MFCD00216472
InChI Key LDKDMDVMMCXTMO-GFCCVEGCSA-N
Synonym n-boc-d-phenylalaninol, boc-d-phenylalaninol, r-tert-butyl 1-hydroxy-3-phenylpropan-2-yl carbamate, n-tert-butoxycarbonyl-d-phenylalaninol, r-2-boc-amino-3-phenyl-1-propanol, r-+-2-tert-butoxycarbonylamino-3-phenyl-1-propanol, r-2-tert-butoxycarbonylamino-3-phenyl-1-propanol, tert-butyl n-2r-1-hydroxy-3-phenylpropan-2-yl carbamate, pubchem5739, r-n-tert-butoxycarbonyl-beta-phenylalaninol
PubChem CID 7019371
IUPAC Name tert-butyl N-[(2R)-1-hydroxy-3-phenylpropan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)N[C@@H](CO)CC1=CC=CC=C1

Specifications

Melting Point 95°C
Color White
Quantity 1 g
Formula Weight 251.33
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-phenylalaninol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.