Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-tert-Butylbenzenesulfenamide 96.0+%, TCI America™
SDP

Catalog No. p-7119575 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g
25 g

Chemical Identifiers

CAS 19117-31-8
Molecular Formula C10H15NS
Molecular Weight (g/mol) 181.30
MDL Number MFCD03844776
InChI Key AAQBFMPKCDTAJD-UHFFFAOYSA-N
Synonym N-tert-Butylphenylsulfenamide
PubChem CID 11182990
IUPAC Name tert-butyl(phenylsulfanyl)amine
SMILES CC(C)(C)NSC1=CC=CC=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 181.30
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material N-tert-Butylbenzenesulfenamide
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.