Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N1-(1-Oxo-2,3-dihydro-1H-inden-5-yl)acetamide, 97%, Thermo Scientific™

Catalog No. TL00757EB
Click to view available options
Quantity:

Chemical Identifiers

CAS

58161-35-6

Molecular Formula

C11H11NO2

Molecular Weight (g/mol)

189.214

MDL Number

MFCD00099465

InChI Key

GHUPGGYDRVSZSW-UHFFFAOYSA-N

Synonym

n-1-oxo-2,3-dihydro-1h-inden-5-yl acetamide, n1-1-oxo-2,3-dihydro-1h-inden-5-yl acetamide, 5-acetamido-1-indanone, 5-acetylamino-1-indanone, n-1-oxoindan-5-yl acetamide, n-1-oxo-2,3-dihydroinden-5-yl acetamide, acetamide, n-2,3-dihydro-1-oxo-1h-inden-5-yl, 5-acetamido-2,3-dihydro-1-oxo-1h-indene, 5-acetamido-indanone

PubChem CID

312894

IUPAC Name

N-(1-oxo-2,3-dihydroinden-5-yl)acetamide

SMILES

CC(=O)NC1=CC2=C(C=C1)C(=O)CC2

Specifications

CAS Min %

97.0

Assay Percent Range

97%

Quantity

25 g

Formula Weight

189.21

Percent Purity

97%

Chemical Name or Material

N1-(1-oxo-2, 3-dihydro-1H-inden-5-yl)acetamide

Hazard Statement

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

Precautionary Statement

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.