Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N2-Isobutyryl-2′-deoxyguanosine 98.0+%, TCI America™
SDP

Catalog No. p-7131551 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 68892-42-2
Molecular Formula C14H19N5O5
Molecular Weight (g/mol) 337.336
MDL Number MFCD00010060
InChI Key SIDXEQFMTMICKG-DJLDLDEBSA-N
PubChem CID 688078
IUPAC Name N-[9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6-oxo-3H-purin-2-yl]-2-methylpropanamide
SMILES CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C3CC(C(O3)CO)O

Specifications

Melting Point 300°C
Color White
Quantity 100 mg
Formula Weight 337.34
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N2-Isobutyryl-2′-deoxyguanosine
TSCA No
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.