Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N2-(tert-Butoxycarbonyl)-4-fluoro-1,2-phenylenediamine 98.0+%, TCI America™
SDP

Catalog No. p-7123421 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 362670-07-3
Molecular Formula C11H15FN2O2
Molecular Weight (g/mol) 226.25
MDL Number MFCD07787180
InChI Key FELBQUPQCJZQQX-UHFFFAOYSA-N
Synonym N2-Boc-4-fluoro-1,2-phenylenediamine, tert-Butyl (2-Amino-5-fluorophenyl)carbamate, (2-Amino-5-fluorophenyl)carbamic Acid tert-Butyl Ester
PubChem CID 53420357
IUPAC Name tert-butyl N-(2-amino-5-fluorophenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=C(N)C=CC(F)=C1

Specifications

Melting Point 101°C
Color Gray-Red
Quantity 200 mg
Formula Weight 226.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N2-(tert-Butoxycarbonyl)-4-fluoro-1,2-phenylenediamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.