Learn More
HARWICK STANDARD DISTRIBUTION N330 Carbon Black
SDP
Supplier:  HARWICK STANDARD DISTRIBUTION N330CARBONBLACK(PELLET,50
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Provides good abrasion resistance with high resilience, easy processing, good tensile and tear properties. Wide range of applications in NR and SR compounds for both tires and mechanical goods.
Catalog No. NC1755360