Learn More
Medchemexpress LLC HY-101854 10mg Medchemexpress, N6-(2-Phenylethyl)adenosine CAS:20125-39-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10185410MG
Encompass_Preferred
Medchemexpress, HY-101854 10mg N6-(2-Phenylethyl)adenosine CAS:20125-39-7 N6-(2-Phenylethyl)adenosine is a selective A1 adenosine receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7889
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.