Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine 98.0+%, TCI America™
SDP

Catalog No. p-7119580 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 2480-93-5
Molecular Formula C18H26N2O6
Molecular Weight (g/mol) 366.414
MDL Number MFCD00038259
InChI Key QYYCZJUFHDLLOJ-AWEZNQCLSA-N
Synonym boc-orn z-oh, s-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, boc-orn cbz-oh, nalpha-boc-ndelta-cbz-l-ornithine, nalpha-tert-butoxycarbonyl-ndelta-carbobenzoxy-l-ornithine, 2s-2-2-methylpropan-2-yl oxycarbonylamino-5-phenylmethoxycarbonylamino pentanoic acid, 2s-5-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, n-tert-butoxycarbonyl-n'-benzyloxycarbonyl-l-ornithine, n-alpha-boc-n-delta-z-l-ornithine, bocnh-orn z-oh
PubChem CID 7269338
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-(phenylmethoxycarbonylamino)pentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1=CC=CC=C1)C(=O)O

Specifications

Melting Point 94°C
Color White-Yellow
Quantity 5 g
Formula Weight 366.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.