Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nanodiamond (particle size : <10nm) (Hydrogen-terminated), TCI America™
SDP

Catalog No. N10571G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g

Specifications

Quantity 1 g
MDL Number MFCD00211867
Physical Form Powder
Chemical Name or Material Nanodiamond (particle size : <10nm) (Hydrogen-terminated)
RTECSNumber HL4158550
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.