Learn More
Thermo Scientific™ NanoDrop™ Lite Spectrophotometer
The Thermo Scientific™ NanoDrop Lite Spectrophotometer is a compact, personal UV-Vis microvolume spectrophotometer that complements the full-featured NanoDrop 2000/2000c and NanoDrop 8000 instruments.
Supplier: Thermo Scientific™ 840281500
Description
New sales of this product have been discontinued. Please see the newest model: NanoDrop Lite Plus Spectrophotometer.
The NanoDrop Lite performs basic microvolume measurements. Its compact design, with built-in controls and software, make the NanoDrop Lite small enought to fit on any benchtop. The patented sample retention system allows sample to be pipetted directly onto the optical measurement surface. After measurement the sample is wiped off the measurement surface with a lint free lab wipe.- Measures nucleic acid concentration at 260nm and purity using the 260/280 ratio
- Measures purified protein concentration at 280nm
- Employs the unique NanoDrop microvolume sampling technology
- Delivers the accuracy and reproducibility expected from NanoDrop instruments
- Uses built-in controls and software–no computer required
- Optional accessory printer for cryogenic labels available
Specifications
0.03 | |
UL/CSA and CE | |
1500 ng/μL (dsDNA); 45 mg/mL (BSA) | |
11.5 cm | |
Silicon Photodiode | |
LED | |
UV-Vis Spectrophotometer with Printer | |
260 to 280 nm | |
Nucleic Acid Quantification, DNA Quantification, RNA Quantification, Protein Quantification | |
303 stainless steel; quartz fiber | |
4 ng/μL (dsDNA); 45 mg/mL (BSA) | |
0.5 mm | |
1 to 2 μL | |
1 μL | |
18 W | |
0.8 kg |
0 to 30 A | |
Local Control Only | |
USB | |
0.12 mg/ml (BSA), 4 ng/μ/ (dsDNA) | |
LCD | |
NanoDrop Lite | |
6 V | |
16 x 11.5 cm | |
Printer | |
<5 sec. | |
1 | |
<8nm | |
8.0 nm | |
1 year parts and labor | |
0.002 (10 individual measurements at 0.74 Abs) | |
16 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.