Learn More
Medicus Health Narcotics Cabinet, Double Door, Double Lock, High Security Keys
Supplier: Medicus Health 2976M1
Narcotics Cabinet, Double Door, Double Lock, High Security Pick Resistant Tubular Locks for high security medication storage regulations that require mandatory double locking protection with two doors and two locks• Double Door, Double Lock, Narcotics Cabinet is intended for safe storage of narcotics, prescriptions, medicine, and other drugs in the pharmacy or hospital• Overall cabinet outside dimensions: 12"W x 8"D x 15"H; Internal cabinet dimensions: 11.5"W x 6"D x 14.5"H• Stainless steel pinned door hinges prevent pin removal• (2) height adjustable shelves• Predrilled holes for mounting on a wall, cabinet or other solid surface (mounting hardware not included)• Narcotics cabinet is delivered ready to use• 2-year warranty against manufacturer defect• Durable hard-baked powder coat finish• Cabinet weighs 15 lbs.• Standard color is beige
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.