Learn More
McKesson General Medical Nasal Cannula Low Flow Delivery Universal Straight Prong / NonFlared Tip
Supplier:  McKesson General Medical 1038444
Encompass_Preferred
Designed for patients requiring medium or low concentrations of oxygen. Individually packaged in poly bags. Recommended flow rate: 1-6 LPM.
Catalog No. NC1715982
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.