Learn More
National Marker™ Chemical Hazard Identification Labels
High visibility red-and-white pressure-sensitive vinyl labels ideal for alerting employees to potential chemical hazards in the workplace
Specifications
Color | White, Red |
---|---|
Format | Label |
Material | Vinyl |
Product Type | Container Label |
For Use With (Application) | For marking larger containers, both indoor and outdoor applications |
Catalog Number | Mfr. No. | Display Text | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Display Text | Price | Quantity & Availability | |||||
17-985-422
|
National Marker™
HC201P |
Acetic Acid | N/A | N/A | N/A | ||||
17-985-423
|
National Marker™
HC200P |
Acetone | N/A | N/A | N/A | ||||
17-985-432
|
National Marker™
HC227P |
Chlorine | N/A | N/A | N/A | ||||
17-985-435
|
National Marker™
HC236P |
Ethyl Alcohol | N/A | N/A | N/A | ||||
17-985-441
|
National Marker™
HC244P |
Formaldehyde | N/A | N/A | N/A | ||||
17-985-439
|
National Marker™
HC241P |
Flammable Liquid | N/A | N/A | N/A | ||||
17-985-451
|
National Marker™
HC260P |
Hydroxide Peroxide | N/A | N/A | N/A | ||||
17-985-446
|
National Marker™
HC248P |
Gasoline | N/A | N/A | N/A | ||||
17-985-452
|
National Marker™
HC262P |
Isopropyl Alcohol | N/A | N/A | N/A | ||||
17-985-456
|
National Marker™
HC269P |
Mercury | N/A | N/A | N/A | ||||
19-035-043
|
National Marker™
HC270P |
Methyl Alcohol | N/A | N/A | N/A | ||||
17-985-464
|
National Marker™
HC281P |
Nitric Acid | N/A | N/A | N/A | ||||
17-985-467
|
National Marker™
HC297P |
Phosphoric Acid | N/A | N/A | N/A | ||||
17-985-472
|
National Marker™
HC307P |
Propane | N/A | N/A | N/A | ||||
17-985-478
|
National Marker™
HC322P |
Toluene | N/A | N/A | N/A | ||||
19-035-039
|
National Marker™
HC330P |
Diesel Fuel | N/A | N/A | N/A | ||||
19-035-046
|
National Marker™
HC345P |
Xylene | N/A | N/A | N/A | ||||
Description
- Made to withstand most chemicals, solvents and vapors
- Resist fading and scratching
- For marking larger containers
- Measure 6 × 10 in.
Specifications
White, Red | |
Vinyl | |
For marking larger containers, both indoor and outdoor applications | |
10 in. | |
15.24 cm |
Label | |
Container Label | |
1 Ea. | |
6 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.