Learn More
Hampton Research NATRIX 2 NO48 185ML
SDP
Supplier:  Hampton Research HR291748
Encompass
HR2-917-48/Natrix 2 #48 (H12), 185 ml - 0.002 M Copper(II) chloride dihydrate, 0.05 M TRIS hydrochloride pH 8.5, 1.8 M Lithium sulfate monohydrate, 0.0005 M Spermine. Please note items from this supplier are non-returnable.
Catalog No. 50-170-3
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.