Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NBI 27914 hydrochloride, Tocris Bioscience™

Catalog No. p-4785210 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective non-peptide CRF1 antagonist

Selective, non-peptide corticotropin-releasing factor1 (CRF1) receptor antagonist (Ki = 1.7 nM); has no activity at CRF2 receptors. Blocks behavioral seizures in vivo.

Chemical Identifiers

CAS 1215766-76-9
Molecular Formula C18H21Cl5N4
Molecular Weight (g/mol) 470.644
InChI Key CPMGENCTAWBLNW-UHFFFAOYSA-N
Synonym nbi 27914 hydrochloride, 5-chloro-n-cyclopropylmethyl-2-methyl-n-propyl-n'-2,4,6-trichlorophenyl-4,6-pyrimidinediamine hydrochloride, 5-chloro-4-n-cyclopropyl methyl-n-propylamino-2-methyl-6-2,4,6-trichlorophenyl aminopyridine
PubChem CID 45073446
IUPAC Name 5-chloro-4-N-(cyclopropylmethyl)-2-methyl-4-N-propyl-6-N-(2,4,6-trichlorophenyl)pyrimidine-4,6-diamine;hydrochloride
SMILES CCCN(CC1CC1)C2=NC(=NC(=C2Cl)NC3=C(C=C(C=C3Cl)Cl)Cl)C.Cl

Specifications

Quantity 10 mg
Formula Weight 470.66
Percent Purity >99%
Chemical Name or Material NBI 27914 hydrochloride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.