Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-NBI 74330, Tocris Bioscience™

Catalog No. 452810 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective CXCR3 antagonist

Potent and selective CXCR3 antagonist; potently inhibits 125I-CXCL10 binding to CXCR3 (pKi = 8.13). Inhibits calcium mobilization in response to CXCL11 and CXCL10 in RBL cells. Exhibits no significant effect on chemotaxis induced by CXCR4 or CCR7. Displays a five-fold greater affinity for CXCR3 than (±)-AMG 487 .

This product is racemic.

Chemical Identifiers

CAS 473722-68-8
Molecular Formula C32H27F4N5O3
Molecular Weight (g/mol) 605.594
InChI Key XMRGQUDUVGRCBS-UHFFFAOYSA-N
Synonym n-1-3-4-ethoxyphenyl-4-oxopyrido 2,3-d pyrimidin-2-yl ethyl-2-4-fluoro-3-trifluoromethyl phenyl-n-pyridin-3-ylmethyl acetamide, d0a8pv, plusmn-nbi 74330, gtpl839, n-1-3-4-ethoxyphenyl-3,4-dihydro-4-oxopyrido 2,3-d pyrimidin-2-yl ethyl-4-fluoro-n-3-pyridinylmethyl-3-trifluoromethyl benzeneacetamide
PubChem CID 10167713
IUPAC Name N-[1-[3-(4-ethoxyphenyl)-4-oxopyrido[2,3-d]pyrimidin-2-yl]ethyl]-2-[4-fluoro-3-(trifluoromethyl)phenyl]-N-(pyridin-3-ylmethyl)acetamide
SMILES CCOC1=CC=C(C=C1)N2C(=O)C3=C(N=CC=C3)N=C2C(C)N(CC4=CN=CC=C4)C(=O)CC5=CC(=C(C=C5)F)C(F)(F)F

Specifications

Quantity 10 mg
Formula Weight 605.58
Chemical Name or Material (±)-NBI 74330
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.