Learn More
New England Biolabs, Inc. NEB Turbo Competent E.coli (High Efficiency) - 20x0.05 ml
SDP
Supplier:  New England Biolabs, Inc. C2984H
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
NEB Turbo Competent E. coli (High Efficiency) - 20x0.05 ml. Chemically competent E. coli cells suitable for high efficiency transformation and rapid colony growth (6.5 hours).Help NEB go green. FREE FREIGHT on NEB orders greater than $350.00!
Catalog No. 50-995-107
missing translation for 'QuantityLeft'