Learn More
New England Biolabs, Inc. NEBuilder HiFi DNA Assembly Master Mix – 250 reactions
SDP
Supplier:  New England Biolabs, Inc. E2621X
Encompass
NEBuilder HiFi DNA Assembly Master Mix was developed to improve the efficiency and accuracy of DNA assembly. It allows for seamless assembly of multiple DNA fragments, regardless of fragment length or end compatibility. Used to assemble either ss oligonucleotides or different sizes of DNA frags with varied overlaps. Has utility in synbio community, as well as those interested in one-step cloning of multiple fragments due to its ease of use, flexibility and simple MM format. The rxn includes different enzymes that work together in the same buffer: The exonuclease creates single-stranded 3' overhangs that facilitate the annealing of fragments that share complementarity at one end (the overlap region): a polymerase fills in gaps within each annealed fragment; DNA ligase seals nicks in the assembled DNA. End result is a ds fully sealed DNA molecule that can serve as template for PCR, RCA or a variety of other molecular biology applications, including direct transformation of E coli.
Catalog No. 50-104-7501
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.