Learn More
New England Biolabs, Inc. NEBuilder HiFi DNA Assembly Master Mix – 10 reactions
SDP
Supplier:  New England Biolabs, Inc. E2621S
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
NEBuilder HiFi DNA Assembly Master Mix was developed to improve the efficiency and accuracy of DNA assembly. It allows for seamless assembly of multiple DNA fragments, regardless of fragment length or end compatibility. Used to assemble either ss oligonucleotides or different sizes of DNA frags with varied overlaps. Has utility in synbio community, as well as those interested in one-step cloning of multiple fragments due to its ease of use, flexibility and simple MM format. The rxn includes different enzymes that work together in the same buffer: The exonuclease creates single-stranded 3' overhangs that facilitate the annealing of fragments that share complementarity at one end (the overlap region): a polymerase fills in gaps within each annealed fragment; DNA ligase seals nicks in the assembled DNA. End result is a ds fully sealed DNA molecule that can serve as template for PCR, RCA or a variety of other molecular biology applications, including direct transformation of E coli.
Catalog No. NC0824638