Learn More
Enzo Life Sciences Necrostatin-1 (100mg). CAS: 4311-88-0
Supplier: Enzo Life Sciences BMLAP3090100
Necrostatin-1 inhibits necroptosis, a non-apoptotic cell death pathway. Inhibits the loss of mitochondrial membrane potential in TNFα-treated Jurkat cells (EC50=490 nM). Does not inhibit FAS-induced apoptosis and has no effect on apoptotic morphology. It displays a pronounced protective effect in a mouse model of ischemic brain injury and inhibits myocardial cell death. Inhibits RIP1 kinase the key upstream kinase involved in the activation of necroptosis (EC50=180nM). Alternative name: Methylthiohydantoin-DL-tryptophan. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (20mg/ml) or 100% ethanol (5mg/ml). Long Term Storage: Ambient.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.