Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Necrostatin-1, Tocris Bioscience™

Catalog No. p-4784185 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

RIP1 kinase inhibitor; inhibits necroptosis

ATP-competitive, allosteric inhibitor of receptor-interacting protein kinase 1 (RIPK1) (EC50 = 182 nM). Blocks non-apoptotic cell death via inhibition of a specific cellular pathway, necroptosis, which leads to necrosis (EC50 = 494 nM). Reduces ischemic brain injury in a mouse model of stroke.

Chemical Identifiers

CAS 4311-88-0
Molecular Formula C13H13N3OS
Molecular Weight (g/mol) 259.327
InChI Key TXUWMXQFNYDOEZ-UHFFFAOYSA-N
Synonym necrostatin-1, mth-dl-tryptophan, nec-1, necrostatin 1, 5-1h-indol-3-ylmethyl-3-methyl-2-thioxo-4-imidazolidinone, mth-trp, methyl-thiohydantoin-tryptophan, necroptotic inhibitor, nec-1, 5-1h-indol-3-ylmethyl-3-methyl-2-sulfanylideneimidazolidin-4-one, 4-imidazolidinone, 5-1h-indol-3-ylmethyl-3-methyl-2-thioxo
PubChem CID 2828334
IUPAC Name 5-(1H-indol-3-ylmethyl)-3-methyl-2-sulfanylideneimidazolidin-4-one
SMILES CN1C(=O)C(NC1=S)CC2=CNC3=CC=CC=C32

Specifications

Quantity 10 mg
Formula Weight 259.33
Percent Purity 98%
Chemical Name or Material Necrostatin-1
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.