Learn More
Medchemexpress LLC HY-104015 10mg Medchemexpress, NecroX-5 CAS:1383718-29-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10401510MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-104015 10mg NecroX-5 CAS:1383718-29-3 NecroX-5 is a derivative of the NecroX, reduces intracellular calcium concentration, and possesses anti-inflammatory and anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8188
missing translation for 'QuantityLeft'