Learn More
Med Vet International Terumo SurGuard Safety Needle with 1cc Syringe, 25G x 5/8", 100/Box
Supplier:  Med Vet International SG301T2516
Encompass_Preferred
Terumo Medical Corp. SG3-1T2516 Safety Needle with 1cc Syringe, 25G x 5/8 in. , 1/box. The industry's most adaptable safety needle with three ways to activate and a broad range of sizes. Add separate thumb and finger grips, unequalled sharpness, as well as, ease of use and simple standardization that make everyone comfortable. Our hypodermic syringes features an ultra-clear barrel and bold scale markings that allow clinicians to quickly confirm the precise dosage. An integrated plunger stop helps prevent the plunger from being accidentally pulled out, and the lack of any natural rubber latex components provides an extra safety measure. Additional Benefits:- 3 modes of activation-thumb, finger, or hard surface- Separate finger/thumb grips with ridges and nonslip surfaces- Hub and needle locks for security post activation- Broad range of sizes- Firm injection feel- Sharpest needles among major brands tested
Catalog No. 50-264-3447
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.