Learn More
Metrohm USA NEEDLE HOLDER FOR KF OVEN
SDP
Supplier:  Metrohm USA 62049050
Encompass
Needle holder (73 mm) with Luer lock for the Karl Fischer ovens 860, 874 and 885.
Catalog No. NC0432402
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.