Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nemonapride, Tocris Bioscience™

Catalog No. p-4784188 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly potent D2-like antagonist. Also 5-HT1A agonist

Highly potent dopamine D2-like receptor antagonist; selective over D1-like receptors (Ki values are 0.1 and 740 nM for D2-like and D1-like receptors respectively). Also potent 5-HT1A receptor agonist (IC50 = 34 nM) and has affinity for sigma receptors.

Chemical Identifiers

CAS 75272-39-8
Molecular Formula C21H26ClN3O2
Molecular Weight (g/mol) 387.908
InChI Key KRVOJOCLBAAKSJ-KSSFIOAISA-N
Synonym nemonapride, --nemonapride, 2s,3s-nemonapride, emonapride, emilace, n-2s,3s-1-benzyl-2-methylpyrrolidin-3-yl-5-chloro-2-methoxy-4-methylamino benzamide, dsstox_cid_22612, dsstox_rid_80057, dsstox_gsid_42612, cis-n-1-benzyl-2-methylpyrrolidin-3-yl-5-chloro-2-methoxy-4-methylaminobenzamide
PubChem CID 9952220
ChEBI CHEBI:64218
IUPAC Name N-[(2S,3S)-1-benzyl-2-methylpyrrolidin-3-yl]-5-chloro-2-methoxy-4-(methylamino)benzamide
SMILES CC1C(CCN1CC2=CC=CC=C2)NC(=O)C3=CC(=C(C=C3OC)NC)Cl

Specifications

Quantity 10 mg
Formula Weight 387.91
Percent Purity >99%
Chemical Name or Material Nemonapride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.