Learn More
TARGETMOL CHEMICALS INC NEOBAVAISOFLAVONE 5MG
Supplier:  TARGETMOL CHEMICALS INC T6S0139
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Neobavaisoflavone is isolated as a DNA polymerase inhibitor. 2. Neobavaisoflavone might be a potential anabolic agent to treat bone loss-associated diseases. 3. Neobavaisoflavone has anti-inflammatory activity can significantly inhibit the production of reactive oxygen species (ROS) reactive nitrogen species (RNS) and cytokines IL-1(beta) IL-6 IL-12p4 IL-12p7 TNF-(alpha) in LPS+IFN-(gamma)- or PMA- stimulated RAW264.7 macrophages. purity: 99%
Catalog No. 50-257-6391
missing translation for 'QuantityLeft'