Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III) acetate hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog No. p-7059428
Click to view available options
Quantity:
50 g
250 g

CAS: 334869-71-5 | C6H9NdO6 | 321.37 g/mol

Neodymium Acetate, mainly used for glass, crystal and capacitors. It is useful in protective lenses for welding goggles. It is also used in CRT displays to enhance contrast between reds and greens. It is highly valued in glass manufacturing for its attractive purple coloring to glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 334869-71-5
Molecular Formula C6H9NdO6
Molecular Weight (g/mol) 321.37
MDL Number MFCD00013040
InChI Key KETUDCKOKOGBJB-UHFFFAOYSA-K
IUPAC Name neodymium(3+) triacetate
SMILES [Nd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Physical Form Crystalline
Quantity 50 g
Solubility Information Soluble in water,moderately soluble in strong mineral acids.
Formula Weight 321.38 (Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Neodymium (III) acetate hydrate
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.