Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III) chloride hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog No. p-4824824
Click to view available options
Quantity:
50 g
250 g

Neodymium(III) chloride hydrate is used for production of neodymium metal. It also finds application in lasers and fiber amplifiers. It serves as dopants for silica based as well as plastic fibers. It plays an important role of additives in organic light emitting diodes resulting in improved LED emission spectrum. In addition, it is used in catalysis of polymerization of various dienes. It is used to modify titanium dioxide for use as a photocatalyst. Its coating on aluminum and its alloy protect them from corrosion. It is also extensively used as fluorescent labels for organic molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19423-78-0
MDL Number MFCD00011132

Specifications

Melting Point 124°C
Physical Form Crystalline Aggregates
Quantity 50 g
Assay Percent Range 99.9% (REO)
Molecular Formula NdCl3·H2O
Solubility Information Soluble in water.
Formula Weight 250.60 (Anhydrous)
Odor Odorless
Appearance Crystalline aggregrates
Chemical Name or Material Neodymium(III) chloride hydrate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-031-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.