Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

Catalog No. p-7049412
Click to view available options
Quantity:
5 g
25 g

CAS: 10024-93-8

Neodymium(III) chloride is used as a precursor for the production of neodymium metal. It is used as a catalyst and accelerates polymerization of various dienes such as polybutylene, polybutadiene, and polyisoprene. It has luminescence property and is widely used as fluorescent label in organic molecules, thereby helping in easy tracking of the compound using fluorescence microscope during various physical and chemical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon

Chemical Identifiers

CAS 10024-93-8
MDL Number MFCD00011132

Specifications

Melting Point 784°C
Physical Form Powder
Quantity 5 g
Merck Index 14,6450
Solubility Information Soluble in water and ethanol.
Formula Weight 250.6
Percent Purity 99.99%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 4.134 g/mL
Assay (REO)
Chemical Name or Material Neodymium(III) chloride, Ultra dry
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-031-5
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.