Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neodymium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog No. p-4408132
Click to view available options
Quantity:
5 g
25 g

It is used as pharmaceutical intermediates. It is used in the selective PVC membrane sensors. The sensor with ionophore (L1) exhibits significantly enhanced selectivity towards neodymium (III).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13746-96-8
Molecular Formula N3NdO9
Molecular Weight (g/mol) 330.25
MDL Number MFCD00149798
InChI Key CFYGEIAZMVFFDE-UHFFFAOYSA-N
Synonym neodymium iii nitrate hydrate, neodymium nitrate hydrate, acmc-20ak53, 3no3.nd.h2o, neodymium nitrate-water 1/3/1, neodymium 3+ ion hydrate trinitrate, neodymium 3+ hydrate trinitrate, neodymium iii nitrate hydrate, reacton, nitric acid,neodymium 3+ salt, hydrate 8ci,9ci
PubChem CID 71311426
SMILES [Nd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point ∽40°C
Color Blue
Physical Form Wet Crystalline Aggregates
Quantity 5 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in water.
IUPAC Name neodymium(3+);trinitrate;hydrate
Formula Weight 330.25 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance Violet crystals
Chemical Name or Material Neodymium(III) nitrate hydrate
Show More Show Less
Hazard Category H272-H315-H319-H335
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 233-153-9
RTECSNumber OJ6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.