Learn More
Neogen Diagnostics K-Gold PNPP Substrate, 500 mL, Microwell, AP, Colorimetric
Supplier:  Neogen Diagnostics 303176
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
K-Gold Substrate is a one-bottle stabilized chromogenic substrate for use with alkaline phosphatase based immunoassays. K-Gold Substrate utilizes the substrate p-nitrophenyl-phosphate (PNPP), which reacts with alkaline phosphatase to produce the chromophore p-nitrophenol. NEOGENs ready-to-use K-Gold formula does not require mixing of additional ingredients or stabilizing agents, making it an ideal formula for kit manufacturers. Stable for a minimum of 30-months when stored at 2-8C.
Catalog No. NC1849076
missing translation for 'QuantityLeft'