Learn More
Gold Biotechnology Inc Neomycin Sulfate, USP Grade 5 g
SDP
Supplier:  Gold Biotechnology Inc N6205
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Neomycin is a broad-spectrum aminoglycoside antibiotic isolated from Streptomyces species. It targets gram-negative and gram-positive bacteria. Neomycin can also be used as a selection agent for prokaryotic cells transformed with the neo gene or NPT II resistance gene and to study toxic antibacterial side effects. In addition, neomycin is a calcium channel protein inhibitor and DNase I inhibitor. It can be used in a variety of cell culture applications.
Catalog No. NC2014280