Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neopentyl Glycol Diacrylate (stabilized with MEHQ) 89.0+%, TCI America™
SDP

Catalog No. p-7133976 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
100 g
500 g

Chemical Identifiers

CAS 2223-82-7
Molecular Formula C11H16O4
Molecular Weight (g/mol) 212.245
MDL Number MFCD00048148
InChI Key MXFQRSUWYYSPOC-UHFFFAOYSA-N
Synonym 2,2-Dimethyl-1,3-propanediol Diacrylate, 1,3-Bis(acryloyloxy)-2,2-dimethylpropane
PubChem CID 16680
IUPAC Name (2,2-dimethyl-3-prop-2-enoyloxypropyl) prop-2-enoate
SMILES CC(C)(COC(=O)C=C)COC(=O)C=C

Specifications

Melting Point 6°C
Color Yellow
Boiling Point 96°C
Quantity 25 g
UN Number 2810
Formula Weight 212.25
Percent Purity ≥89.0% (GC)
Physical Form Liquid
Chemical Name or Material Neopentyl Glycol Diacrylate (stabilized with MEHQ)
EINECSNumber (2)-1007&(2)-0958
RTECSNumber AS8925000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.