Learn More
Enzo Life Sciences Netrin-1 (mouse), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX522124C010
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Concentration: 0.1mg/ml after reconstitution. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of mouse netrin-1 (aa 27-604) is fused at the C-terminus to a FLAG-tag. Specificity: Binds to human and mouse UNC5B. Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-200-9439
missing translation for 'QuantityLeft'