Learn More
CPC Scientific H-Lys-Ile-Pro-Tyr-Ile-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP037A
Encompass
Endogenous neurotensin-like neuropeptide from porcine spinal cord that exhibits contractile activity on guinea-pig ileum and induces hypotension in rats. It also acts as a potent modulator of dopamine D2 receptor agonist binding.SEQUENCE: H-Lys-Ile-Pro-Tyr-Ile-Leu-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: KIPYILMOLECULAR FORMULA: C38H63N7O8MOLECULAR WEIGHT: 745.96STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [92169-45-4]SYNONYMS: RESEARCH AREA: NeuropeptidesREFERENCES:N. Minamino et al., BBRC, 122, 542 (1984)SKU(s): NEUP-037A
Catalog No. 50-210-9922
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.